missing translation for 'onlineSavingsMsg'
Learn More

2-Aminofluorene, 98%

CAS: 153-78-6 | C13H11N | 181.238 g/mol

$360.12 - $360.12

Chemical Identifiers

CAS 153-78-6
Molecular Formula C13H11N
Molecular Weight (g/mol) 181.238
MDL Number MFCD00001125
InChI Key CFRFHWQYWJMEJN-UHFFFAOYSA-N
Synonym 2-aminofluorene, 2-fluorenamine, aminofluoren, 2-fluoreneamine, fluorene, 2-amino, fluoren-2-amine, 2-fluorenylamine, 2-fluroenylamine, fluoren-2-ylamine, aminofluoren german
PubChem CID 1539
IUPAC Name 9H-fluoren-2-amine
SMILES C1C2=CC=CC=C2C3=C1C=C(C=C3)N
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2276914
View Documents
Thermo Scientific Chemicals
B2276914
25 g
Each for $360.12
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

153-78-6
181.238
CFRFHWQYWJMEJN-UHFFFAOYSA-N
1539
C1C2=CC=CC=C2C3=C1C=C(C=C3)N
C13H11N
MFCD00001125
2-aminofluorene, 2-fluorenamine, aminofluoren, 2-fluoreneamine, fluorene, 2-amino, fluoren-2-amine, 2-fluorenylamine, 2-fluroenylamine, fluoren-2-ylamine, aminofluoren german
9H-fluoren-2-amine

Specifications

153-78-6
C13H11N
25 g
2-aminofluorene, 2-fluorenamine, aminofluoren, 2-fluoreneamine, fluorene, 2-amino, fluoren-2-amine, 2-fluorenylamine, 2-fluroenylamine, fluoren-2-ylamine, aminofluoren german
C1C2=CC=CC=C2C3=C1C=C(C=C3)N
181.238
181.24
2-Aminofluorene
126°C to 130°C
MFCD00001125
1945861
CFRFHWQYWJMEJN-UHFFFAOYSA-N
9H-fluoren-2-amine
1539
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H341-H351
Suspected of causing genetic defects.
Suspected of causing cancer.

P201-P202-P280-P308+P313-P501c

H341-H351

EINECSNumber : 205-817-8

RTECSNumber : LL5075000

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only