missing translation for 'onlineSavingsMsg'
Learn More

2,3-O-Isopropylidene-D-erythronolactone, 98%, Thermo Scientific Chemicals

For use in spiroannulated carbohydrate synthesis

Supplier:  Thermo Scientific Chemicals J6687106

 View more versions of this product

Catalog No. AAJ6687106


Only null left
Explore available promotions

Description

Description

It undergoes Aldol condensations with silyl ketene acetals.1 Employed in spiroannulated carbohydrate synthesis.2 Convergent syntheses of a hydroxylated indolizidine,3 carbohydrate substituted benzoquinones,4 and of the oxazole segment of calyculin5 have been accomplished using this chiral synthon.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It undergoes Aldol condensations with silyl ketene acetals.1 Employed in spiroannulated carbohydrate synthesis.2 Convergent syntheses of a hydroxylated indolizidine,3 carbohydrate substituted benzoquinones,4 and of the oxazole segment of calyculin5 have been accomplished using this chiral synthon.

Solubility
Soluble in water

Notes
Store at room temperature. Incompatible with heat, oxidizing agents.
Greener Choice Claims
  • Environmental benefits include:
    • Recyclable
Specifications
Show More
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only