missing translation for 'onlineSavingsMsg'
Learn More

(1R,2R)-(-)-1,2-Diaminocyclohexane, 98%

CAS: 20439-47-8 | C6H14N2 | 114.19 g/mol

Supplier:  Thermo Scientific Chemicals L1405903

 View more versions of this product

Catalog No. AAL1405903


Only null left
Explore available promotions

Description

Description

(1R,2R)-(-)-1,2-Diaminocyclohexane is a versatile ligand for the formation of metal complexes. Used in the synthesis of chiral tropocoronands which have potential utility in asymmetric catalysis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(1R,2R)-(-)-1,2-Diaminocyclohexane is a versatile ligand for the formation of metal complexes. Used in the synthesis of chiral tropocoronands which have potential utility in asymmetric catalysis.

Solubility
Soluble in water.

Notes
Air sensitive. Store under inert gas. Store away from oxidizing agents, air, carbondioxide.
Greener Choice Claims
  • Environmental benefits include:
    • Renewable Energy
Specifications
Show More
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H317
Causes severe skin burns and eye damage.
Causes serious eye damage.
May cause an allergic skin reaction.

P261-P264b-P271-P272-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P333+P313-P363-P501c

H314-H317-H318-H335

DOTInformation : Hazard Class: 8; Packaging Group: II

TSCA : No

Recommended Storage : Keep cold

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only