missing translation for 'onlineSavingsMsg'
Learn More

(1R,2R)-(-)-1,2-Diaminocyclohexane, 98%

Catalog No. p-7046357
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1405903 1 g
AAL1405906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1405903 Supplier Thermo Scientific Chemicals Supplier No. L1405903

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 20439-47-8 | C6H14N2 | 114.19 g/mol

(1R,2R)-(-)-1,2-Diaminocyclohexane is a versatile ligand for the formation of metal complexes. Used in the synthesis of chiral tropocoronands which have potential utility in asymmetric catalysis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(1R,2R)-(-)-1,2-Diaminocyclohexane is a versatile ligand for the formation of metal complexes. Used in the synthesis of chiral tropocoronands which have potential utility in asymmetric catalysis.

Solubility
Soluble in water.

Notes
Air sensitive. Store under inert gas. Store away from oxidizing agents, air, carbondioxide.

Chemical Identifiers

CAS 20439-47-8
Molecular Formula C6H14N2
Molecular Weight (g/mol) 114.19
MDL Number MFCD00062985,MFCD00063747
InChI Key SSJXIUAHEKJCMH-UHFFFAOYNA-N
Synonym 1r,2r---1,2-diaminocyclohexane, 1r,2r-cyclohexane-1,2-diamine, trans-1,2-diaminocyclohexane, trans-1,2-cyclohexanediamine, trans-cyclohexane-1,2-diamine, 1r,2r-1,2-diaminocyclohexane, 1r,2r---1,2-cyclohexanediamine, 1r,2r-diaminocyclohexane, r-dach, 1r,2r-1,2-cyclohexanediamine
PubChem CID 43806
SMILES NC1CCCCC1N

Specifications

Melting Point 40°C to 43°C
Boiling Point 104°C to 110°C (40 mmHg)
Flash Point 70°C (158°F)
Odor Amine-like
Quantity 1 g
UN Number UN3259
Beilstein 4780911
Sensitivity Air sensitive
Solubility Information Soluble in water.
Optical Rotation −25.5° (c=5 in 1N HCl)
IUPAC Name (1R,2R)-cyclohexane-1,2-diamine
Formula Weight 114.19
Percent Purity 98%
Chemical Name or Material (1R,2R)-(-)-1,2-Diaminocyclohexane
Show More Show Less
Hazard Category H314-H317-H318-H335
Hazard Statement GHS H Statement
H314-H318-H317
Causes severe skin burns and eye damage.
Causes serious eye damage.
May cause an allergic skin reaction.
Precautionary Statement P261-P264b-P271-P272-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P333+P313-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: II
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only