Learn More
1H-[1,2,4]Oxadiazolo[4,3-a]-quinoxalin-1-one, 98+%
Selective inhibitor of nitric oxide-sensitive guanylyl cyclase | CAS: 41443-28-1 | C9H5N3O2 | 187.158 g/mol
Supplier: Thermo Scientific Chemicals J62250MF
Description
Selective inhibitor of nitric oxide-sensitive guanylyl cyclase
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 1H-[1,2,4]Oxadiazolo[4,3-a]-quinoxalin-1-one | |
| 160°C to 170°C | |
| C9H5N3O2 | |
| 50 mg | |
| LZMHWZHOZLVYDL-UHFFFAOYSA-N | |
| [1,2,4]oxadiazolo[4,3-a]quinoxalin-1-one | |
| 1456 | |
| ≥99% |
| 41443-28-1 | |
| White | |
| MFCD00792620 | |
| 1h-1,2,4 oxadiazolo 4,3-a quinoxalin-1-one, odq, 1,2,4 oxadiazolo 4,3-a quinoxalin-1-one, unii-s57v2nmv38, 1h-1,2,4 oxadiazolo 4,3-a-quinoxalin-1-one, 1h-1,2,4 oxadiazolo-4,3,a quinoxalin-1-one, 1-h-1,2,4 oxadiazolo-4,3,2 quinoxalin-1-ine, 1h-odq | |
| C1=CC=C2C(=C1)N=CC3=NOC(=O)N23 | |
| 187.158 | |
| 187.15 | |
| Solid |
Chemical Identifiers
| 41443-28-1 | |
| 187.158 | |
| LZMHWZHOZLVYDL-UHFFFAOYSA-N | |
| 1456 | |
| C1=CC=C2C(=C1)N=CC3=NOC(=O)N23 |
| C9H5N3O2 | |
| MFCD00792620 | |
| 1h-1,2,4 oxadiazolo 4,3-a quinoxalin-1-one, odq, 1,2,4 oxadiazolo 4,3-a quinoxalin-1-one, unii-s57v2nmv38, 1h-1,2,4 oxadiazolo 4,3-a-quinoxalin-1-one, 1h-1,2,4 oxadiazolo-4,3,a quinoxalin-1-one, 1-h-1,2,4 oxadiazolo-4,3,2 quinoxalin-1-ine, 1h-odq | |
| [1,2,4]oxadiazolo[4,3-a]quinoxalin-1-one |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
RTECSNumber : RO0913150
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only