missing translation for 'onlineSavingsMsg'
Learn More

1H-[1,2,4]Oxadiazolo[4,3-a]-quinoxalin-1-one, 98+%

Catalog No. p-7042354
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ62250MA 10 mg
AAJ62250MF 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ62250MA Supplier Thermo Scientific Chemicals Supplier No. J62250MA
Only null left

Selective inhibitor of nitric oxide-sensitive guanylyl cyclase | CAS: 41443-28-1 | C9H5N3O2 | 187.158 g/mol

Selective inhibitor of nitric oxide-sensitive guanylyl cyclase

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 41443-28-1
Molecular Formula C9H5N3O2
Molecular Weight (g/mol) 187.158
MDL Number MFCD00792620
InChI Key LZMHWZHOZLVYDL-UHFFFAOYSA-N
Synonym 1h-1,2,4 oxadiazolo 4,3-a quinoxalin-1-one, odq, 1,2,4 oxadiazolo 4,3-a quinoxalin-1-one, unii-s57v2nmv38, 1h-1,2,4 oxadiazolo 4,3-a-quinoxalin-1-one, 1h-1,2,4 oxadiazolo-4,3,a quinoxalin-1-one, 1-h-1,2,4 oxadiazolo-4,3,2 quinoxalin-1-ine, 1h-odq
PubChem CID 1456
IUPAC Name [1,2,4]oxadiazolo[4,3-a]quinoxalin-1-one
SMILES C1=CC=C2C(=C1)N=CC3=NOC(=O)N23

Specifications

Melting Point 160°C to 170°C
Color White
Quantity 10 mg
Formula Weight 187.15
Percent Purity ≥99%
Physical Form Solid
Chemical Name or Material 1H-[1,2,4]Oxadiazolo[4,3-a]-quinoxalin-1-one
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
RTECSNumber RO0913150
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only