missing translation for 'onlineSavingsMsg'
Learn More

1-Phenyl-1-hexyn-3-ol, 97%

CAS: 1817-51-2 | C12H14O | 174.243 g/mol

Supplier:  Thermo Scientific Chemicals H5343714

 View more versions of this product

Catalog No. AAH5343714

Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Greener Choice Claims
  • Environmental benefits include:
    • Recyclable
Specifications
Show More
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only