missing translation for 'onlineSavingsMsg'
Learn More

1-Phenyl-1-hexyn-3-ol, 97%

Catalog No. p-7035712
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5343706 5 g
AAH5343714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5343706 Supplier Thermo Scientific Chemicals Supplier No. H5343706

CAS: 1817-51-2 | C12H14O | 174.243 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1817-51-2
Molecular Formula C12H14O
Molecular Weight (g/mol) 174.243
MDL Number MFCD00046687
InChI Key HGUIQUVFOYTZNC-UHFFFAOYSA-N
Synonym 1-phenyl-1-hexyn-3-ol, 1-phenyl-1-hexyne-3-ol, 1-hexyn-3-ol, 1-phenyl
PubChem CID 582987
IUPAC Name 1-phenylhex-1-yn-3-ol
SMILES CCCC(C#CC1=CC=CC=C1)O

Specifications

Density 0.949
Flash Point 110°C (230°F)
Refractive Index 1.549
Quantity 5 g
Formula Weight 174.24
Percent Purity 97%
Chemical Name or Material 1-Phenyl-1-hexyn-3-ol
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only