Learn More
1,4-Dibromobenzene, 99%
CAS: 106-37-6 | C6H4Br2 | 235.91 g/mol
Supplier: Thermo Scientific Chemicals 112730010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Environmental benefits include:
- Renewable Energy
Specifications
1, 4-Dibromobenzene | |
98.5 | |
86°C to 89°C | |
1.8400g/mL | |
99% | |
C6H4Br2 | |
MFCD00000089 | |
05, 211 | |
p-dibromobenzene, p-bromophenyl bromide, benzene, 1,4-dibromo, benzene, p-dibromo, p-benzene dibromide, 1,4-dibromo benzene, 1,4-dibromo-benzene, unii-9991w3m5hz, dibromobenzene, 1,4, labotest-bb ltbb002071 | |
BrC1=CC=C(Br)C=C1 | |
235.91 | |
CHEBI:37150 | |
≥98.5% |
106-37-6 | |
100.0 | |
Colorless to White | |
Authentic | |
Plastic bottle | |
C6H4Br2 | |
1 kg | |
15, 3024 | |
SWJPEBQEEAHIGZ-UHFFFAOYSA-N | |
1,4-dibromobenzene | |
7804 | |
235.91 | |
Crystals |
Chemical Identifiers
106-37-6 | |
235.91 | |
SWJPEBQEEAHIGZ-UHFFFAOYSA-N | |
7804 | |
1,4-dibromobenzene |
C6H4Br2 | |
MFCD00000089 | |
p-dibromobenzene, p-bromophenyl bromide, benzene, 1,4-dibromo, benzene, p-dibromo, p-benzene dibromide, 1,4-dibromo benzene, 1,4-dibromo-benzene, unii-9991w3m5hz, dibromobenzene, 1,4, labotest-bb ltbb002071Show More | |
CHEBI:37150 | |
BrC1=CC=C(Br)C=C1 |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Toxic to aquatic life with long lasting effects.
Causes skin irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
RUO – Research Use Only