Learn More
(-)-1,2-Bis((2R,5R)-2,5-diphenylphospholano)ethane, 95%
CAS: 528565-79-9 | C34H36P2 | 506.61 g/mol
Supplier: Thermo Scientific Chemicals H6004603
Description
It is used as ligands like DuPhos and BPE ligands and are highly efficient privileged ligands.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is used as ligands like DuPhos and BPE ligands and are highly efficient privileged ligands.
Solubility
Sparingly soluble in water.(3.2E-5Â g/L) (25°C
Notes
Store at room temperature. Keep away from oxidizing agents.
- Environmental benefits include:
- Renewable Energy
Specifications
1,2-Bis[(2R,5R)-2,5-diphenyl-1-phospholanyl]ethane | |
143°C to 147°C | |
MFCD07369027 | |
Air Sensitive | |
Sparingly soluble in water.(3.2E-5 g/L) (25°C | |
C(CP1[C@H](CC[C@@H]1C1=CC=CC=C1)C1=CC=CC=C1)P1[C@H](CC[C@@H]1C1=CC=CC=C1)C1=CC=CC=C1 | |
506.61 | |
506.6 |
528565-79-9 | |
C34H36P2 | |
1 g | |
--1,2-bis 2r,5r-2,5-diphenylphospholano ethane, --1,2-bis 2r,5r-2,5-diphenylphospholano ethane,, 1,2-bis 2r,5r-2,5-diphenylphospholan-1-yl ethane, 1,1'-ethylenebis 2beta,5alpha-diphenyltetrahydro-1h-phosphole, --1,2-bis 2r,5r-2,5-diphenylphospholano ethane, kanata purity, 2r,5r-1-2-2r,5r-2,5-diphenylphospholan-1-yl ethyl-2,5-diphenylphospholane | |
VHHAZLMVLLIMHT-DXCZPEQUNA-N | |
(2R,5R)-1-[2-[(2R,5R)-2,5-diphenylphospholan-1-yl]ethyl]-2,5-diphenylphospholane | |
11420783 | |
≥97% |
Chemical Identifiers
528565-79-9 | |
506.61 | |
VHHAZLMVLLIMHT-DXCZPEQUNA-N | |
11420783 |
C34H36P2 | |
MFCD07369027 | |
--1,2-bis 2r,5r-2,5-diphenylphospholano ethane, --1,2-bis 2r,5r-2,5-diphenylphospholano ethane,, 1,2-bis 2r,5r-2,5-diphenylphospholan-1-yl ethane, 1,1'-ethylenebis 2beta,5alpha-diphenyltetrahydro-1h-phosphole, --1,2-bis 2r,5r-2,5-diphenylphospholano ethane, kanata purity, 2r,5r-1-2-2r,5r-2,5-diphenylphospholan-1-yl ethyl-2,5-diphenylphospholane | |
C(CP1[C@H](CC[C@@H]1C1=CC=CC=C1)C1=CC=CC=C1)P1[C@H](CC[C@@H]1C1=CC=CC=C1)C1=CC=CC=C1 |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only