missing translation for 'onlineSavingsMsg'
Learn More

(-)-1,2-Bis((2R,5R)-2,5-diphenylphospholano)ethane, 95%

Catalog No. p-7040300
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH60046MD 250 mg
AAH6004603 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH60046MD Supplier Thermo Scientific Chemicals Supplier No. H60046MD
Only null left

CAS: 528565-79-9 | C34H36P2 | 506.61 g/mol

It is used as ligands like DuPhos and BPE ligands and are highly efficient privileged ligands.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as ligands like DuPhos and BPE ligands and are highly efficient privileged ligands.

Solubility
Sparingly soluble in water.(3.2E-5 g/L) (25°C

Notes
Store at room temperature. Keep away from oxidizing agents.

Chemical Identifiers

CAS 528565-79-9
Molecular Formula C34H36P2
Molecular Weight (g/mol) 506.61
MDL Number MFCD07369027
InChI Key VHHAZLMVLLIMHT-DXCZPEQUNA-N
Synonym --1,2-bis 2r,5r-2,5-diphenylphospholano ethane, --1,2-bis 2r,5r-2,5-diphenylphospholano ethane,, 1,2-bis 2r,5r-2,5-diphenylphospholan-1-yl ethane, 1,1'-ethylenebis 2beta,5alpha-diphenyltetrahydro-1h-phosphole, --1,2-bis 2r,5r-2,5-diphenylphospholano ethane, kanata purity, 2r,5r-1-2-2r,5r-2,5-diphenylphospholan-1-yl ethyl-2,5-diphenylphospholane
PubChem CID 11420783
SMILES C(CP1[C@H](CC[C@@H]1C1=CC=CC=C1)C1=CC=CC=C1)P1[C@H](CC[C@@H]1C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 143°C to 147°C
Quantity 250 mg
Sensitivity Air Sensitive
Solubility Information Sparingly soluble in water.(3.2E-5 g/L) (25°C
IUPAC Name (2R,5R)-1-{2-[(2R,5R)-2,5-diphenylphospholan-1-yl]ethyl}-2,5-diphenylphospholane
Formula Weight 506.6
Percent Purity ≥97%
Chemical Name or Material 1,2-Bis[(2R,5R)-2,5-diphenyl-1-phospholanyl]ethane
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only