Get a Microvolume Plate with Agilent BioTek Epoch 2 Purchase
New Lab Start-up Program
When you purchase a qualifying Agilent BioTek Epoch 2 Microplate Spectrophotometer, you’ll get a Take3 Microvolume Plate at no additional cost. To take advantage of this offer, order using the catalog numbers below, log in to the Redemption Center, and complete the redemption form to receive your microvolume plate.
NOTE: Promotion may be used for up to two qualifying products purchased on the same order.
Features and Benefits
The Agilent BioTek Epoch 2 Microplate Spectrophotometer performs UV-Vis absorbance measurements in 6- to 384-well microplates, cuvettes, and in microvolume samples. The Take3 Microvolume Plate lets you decrease sample size to a 2 μL volume. Each Take3 plate includes spaces for 16 x 2 μL microspots, one standard 1 cm cuvette, and two BioCells.
Get a Take3 Plate with Agilent BioTek Epoch 2 Purchase
Redeem Now
Promotion: 649
You must apply and qualify for NLSU to redeem this promotion.
Qualifying Products
    Catalog Number Description Price Quantity & Availability  
Success
BTEPOCH2NSC Details
GREENER_CHOICE
Agilent BioTek Epoch 2 Microplate Spectrophotometer Includes: Cuvette Port
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Product Specifications
Wavelength Range 200 to 999 nm, 1 nm increments
Includes Cuvette Port
Height (English) 22 in.
Measurement Speed 96 wells: 8 sec., 384 wells: 14 sec.
Model EPOCH2NSC
For Use With (Application) Nucleic acid and protein quantification to ELISA and microbial growth assays
Light Source Xenon Flash Lamp
Hertz 50/60 Hz
Certifications/Compliance CE and TUV marked. RoHS compliant.
Accuracy ±2 nm
Detector Type Photodiode
BTEPOCH2TSC Details
GREENER_CHOICE
Agilent BioTek Epoch 2 Microplate Spectrophotometer Includes: Touchscreen; Cuvette port
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Product Specifications
Wavelength Range 200 to 999 nm, 1 nm increments
Includes Touchscreen; Cuvette port
Height (English) 22 in.
Measurement Speed 96 wells: 8 sec., 384 wells: 14 sec.
Model EPOCH2TSC
For Use With (Application) Nucleic acid and protein quantification to ELISA and microbial growth assays
Light Source Xenon Flash Lamp
Hertz 50/60 Hz
Certifications/Compliance CE and TUV marked. RoHS compliant.
Accuracy ±2 nm
Detector Type Photodiode
BTEPOCH2TSI Details
GREENER_CHOICE
Agilent BioTek Epoch 2 Microplate Spectrophotometer, IVD Includes: Touchscreen only
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Product Specifications
Wavelength Range 200 to 999 nm, 1 nm increments
Includes Touchscreen only
Height (English) 22 in.
Measurement Speed 96 wells: 8 sec., 384 wells: 14 sec.
Model EPOCH2TS
For Use With (Application) Nucleic acid and protein quantification to ELISA and microbial growth assays
Light Source Xenon Flash Lamp
Hertz 50/60 Hz
Certifications/Compliance CE and TUV marked. RoHS compliant.
Accuracy ±2 nm
Detector Type Photodiode
BTEPOCH2NCI Details
GREENER_CHOICE
Agilent BioTek Epoch 2 Microplate Spectrophotometer, IVD Includes: Cuvet port only
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Product Specifications
Wavelength Range 200 to 999 nm, 1 nm increments
Includes Cuvet port only
Height (English) 22 in.
Measurement Speed 96 wells: 8 sec., 384 wells: 14 sec.
Model EPOCH2NSC
For Use With (Application) Nucleic acid and protein quantification to ELISA and microbial growth assays
Light Source Xenon Flash Lamp
Hertz 50/60 Hz
Certifications/Compliance CE and TUV marked. RoHS compliant.
Accuracy ±2 nm
Detector Type Photodiode
BTEPOCH2NSI Details
GREENER_CHOICE
Agilent BioTek Epoch 2 Microplate Spectrophotometer, IVD Includes: No Touchscreen; No cuvette port
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Product Specifications
Wavelength Range 200 to 999 nm, 1 nm increments
Includes No Touchscreen; No cuvette port
Height (English) 22 in.
Measurement Speed 96 wells: 8 sec., 384 wells: 14 sec.
Model EPOCH2NS
For Use With (Application) Nucleic acid and protein quantification to ELISA and microbial growth assays
Light Source Xenon Flash Lamp
Hertz 50/60 Hz
Certifications/Compliance CE and TUV marked. RoHS compliant.
Accuracy ±2 nm
Detector Type Photodiode
BTEPOCH2TCI Details
GREENER_CHOICE
Agilent BioTek Epoch 2 Microplate Spectrophotometer, IVD Includes: Touchscreen; Cuvette port
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Product Specifications
Wavelength Range 200 to 999 nm, 1 nm increments
Includes Touchscreen; Cuvette port
Height (English) 22 in.
Measurement Speed 96 wells: 8 sec., 384 wells: 14 sec.
Model EPOCH2TSC
For Use With (Application) Nucleic acid and protein quantification to ELISA and microbial growth assays
Light Source Xenon Flash Lamp
Hertz 50/60 Hz
Certifications/Compliance CE and TUV marked. RoHS compliant.
Accuracy ±2 nm
Detector Type Photodiode
BTEPOCH2TS Details
GREENER_CHOICE
Agilent BioTek Epoch 2 Microplate Spectrophotometer Includes: Touchscreen only
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Product Specifications
Wavelength Range 200 to 999 nm, 1 nm increments
Includes Touchscreen only
Height (English) 22 in.
Measurement Speed 96 wells: 8 sec., 384 wells: 14 sec.
Model EPOCH2TS
For Use With (Application) Nucleic acid and protein quantification to ELISA and microbial growth assays
Light Source Xenon Flash Lamp
Hertz 50/60 Hz
Certifications/Compliance CE and TUV marked. RoHS compliant.
Accuracy ±2 nm
Detector Type Photodiode
BTEPOCH2NS Details
GREENER_CHOICE
Agilent BioTek Epoch 2 Microplate Spectrophotometer Includes: No Touchscreen; No cuvette port
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Product Specifications
Wavelength Range 200 to 999 nm, 1 nm increments
Includes No Touchscreen; No cuvette port
Height (English) 22 in.
Measurement Speed 96 wells: 8 sec., 384 wells: 14 sec.
Model EPOCH2NS
For Use With (Application) Nucleic acid and protein quantification to ELISA and microbial growth assays
Light Source Xenon Flash Lamp
Hertz 50/60 Hz
Certifications/Compliance CE and TUV marked. RoHS compliant.
Accuracy ±2 nm
Detector Type Photodiode
Redeemable Product
Catalog NumberDescription
Catalog Number
Description
Agilent BioTek Take3 Microvolume Plate
Learn More
Restrictions
Offer valid from July 1, 2023 to June 30, 2024. Redemptions must be received by June 30, 2025.
Redeemed product value is based on Fisher Scientific list price. The total value of redemption items cannot exceed the maximum redeemable threshold of the promotion. Offer can be redeemed one time per new laboratory. Offer cannot be combined with any other offers. Orders cannot be combined to qualify unless otherwise stated. Each qualifying item can only count towards one promotion redemption.
Promotion may be used for up to two qualifying products purchased on the same order.