Learn More
Agilent BioTek Take3 Microvolume Plate
GREENER_CHOICE
Catalog No. BTTAKE3SN
Change view
Click to view available options
For Use With (Equipment):
Microplate Reader
Catalog No. For Use With (Equipment)
BTTAKE3SN Microplate Reader
1 options
Catalog No. BTTAKE3SN Supplier Agilent BioTek Supplier No. TAKE3SN
  • ONLINE EXCLUSIVE PRICE
    $5,752.92
    $4,492.34 / Each
    Save $1,260.58
    22.0% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
The Take3 microvolume plate increases the functionality of your microplate reader by decreasing the required sample size down to a 2 μL volume. Each plate is compatible with the Cytation, Synergy, and Epoch readers for microvolume absorbance and fluorescence quantification.
Microvolume quantification is fast and easy by measuring multiple 2 μL samples at the same time, without diluting and without the need for specialized equipment. Preprogrammed nucleic acid and protein protocols in the Take3 app bring confidence in your quantification data.
  • Low volume nucleic acid and protein quantification
    • It takes only 2 μL of undiluted sample to get rapid results with the Take3 plate. Take3 plates are compatible with most Agilent BioTek multimode readers, imaging multimode readers, and microplate spectrophotometers.
  • Easy to clean and add more samples
    • Take3 plates are so easy to clean – simply wipe samples off after measurement with a laboratory wipe.
  • Read 2 μL microspots, BioCells, or standard cuvettes
    • Along with 16 microspots, the Take3 plate can accept a standard 1 cm cuvette or 2 patented BioCells for rapid 1 cm measurements in an Agilent BioTek microplate spectrophotometer.
  • Take3 App
    • Preprogrammed protocols in the Take3 app for dsDNA, ssDNA, RNA, and protein (BSA, IgG, lysozyme) measurement make getting direct quantitative results quick and easy. Rapid visual results are output in convenient tab delimited .txt formats.
  • Instrument compatibility: Cytation C10, Cytation 7, Cytation 5, Cytation 1, Synergy Neo2, Synergy H1, Synergy HTX, Synergy LX, Epoch 2, Epoch
Specifications
Product Type Microvolume Plate
For Use With (Equipment) Microplate Reader
Plate Type 16 x 2 μL microspots, 1 standard 1 cm cuvette and 2 BioCells