accessibility menu, dialog, popup

UserName

Enjoy Up to 50% Off Spotlight Deals All Year Long

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Main Menu
Documents and Certificates

CAS RN 927-21-9

H3CNH2OHNONHOHO

CAS RN 927-21-9

IUPAC Name: 2-[2-(2-aminopropanamido)acetamido]acetic acid
Synonyms: [2-(2-aminopropanamido)acetamido]acetic acid
Molecular Weight (g/mol): 203.2
Molecular Formula: C7H13N3O4
InChi Key: VGPWRRFOPXVGOH-UHFFFAOYNA-N
SMILES: CC(N)C(=O)NCC(=O)NCC(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.