missing translation for 'onlineSavingsMsg'
Learn More

DL-Alanylglycylglycine Dihydrate 98.0+%, TCI America™

Supplier:  TCI America A0184100MG

Catalog No. A0184100MG


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

≥98.0% (T)
White
Crystal-Powder at 20°C
DL-Alanylglycylglycine Dihydrate
927-21-9
Refrigerator
221.213
100mg
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions