accessibility menu, dialog, popup

UserName

Enjoy Up to 50% Off Spotlight Deals All Year Long

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Main Menu
Documents and Certificates

CAS RN 19099-54-8

H3CCH3I

CAS RN 19099-54-8

IUPAC Name: 1-iodo-2-(propan-2-yl)benzene
Synonyms: 1-iodo-2-isopropylbenzene
Molecular Weight (g/mol): 246.09
Molecular Formula: C9H11I
InChi Key: SORQIYFSJAWBNQ-UHFFFAOYSA-N
SMILES: CC(C)C1=CC=CC=C1I

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.