CAS RN 151-21-3
Sodium Lauryl Sulfate (Powder/NF/FCC), Fisher Chemical™
CAS: 151-21-3 Molecular Formula: C12H25NaO4S Molecular Weight (g/mol): 288.38 MDL Number: MFCD00036175 InChI Key: DBMJMQXJHONAFJ-UHFFFAOYSA-M Synonym: Sodium Dodecyl Sulfate,SDS,Dodecylsulfuric Acid Sodium Salt,Duponol™ C Dry PubChem CID: 3423265 ChEBI: CHEBI:8984 IUPAC Name: sodium dodecyl sulfate SMILES: [Na+].CCCCCCCCCCCCOS([O-])(=O)=O
Sodium Dodecyl Sulfate (SDS), White Powder, Electrophoresis, Fisher BioReagents™
Molecular Formula: C12H25NaO4S Molecular Weight (g/mol): 288.38 MDL Number: MFCD00036175 InChI Key: DBMJMQXJHONAFJ-UHFFFAOYSA-M Synonym: Sodium Lauryl Sulfate,SDS PubChem CID: 3423265 ChEBI: CHEBI:8984 IUPAC Name: sodium dodecyl sulfate SMILES: [Na+].CCCCCCCCCCCCOS([O-])(=O)=O
Sodium Dodecyl Sulfate (SDS), Micropellets, Fisher BioReagents
CAS: 151-21-3 Molecular Formula: C12H25NaO4S Molecular Weight (g/mol): 288.38 MDL Number: MFCD00036175 InChI Key: DBMJMQXJHONAFJ-UHFFFAOYSA-M PubChem CID: 3423265 ChEBI: CHEBI:8984 SMILES: [Na+].CCCCCCCCCCCCOS([O-])(=O)=O
SDS Solution, 20% Sodium Dodecyl Sulfate Solution, Molecular Biology/Electrophoresis, Fisher BioReagents™
CAS: 151-21-3 Molecular Formula: C12H25NaO4S Molecular Weight (g/mol): 288.38 MDL Number: MFCD00036175 InChI Key: DBMJMQXJHONAFJ-UHFFFAOYSA-M Synonym: Sodium lauryl sulfate,SDS PubChem CID: 3423265 ChEBI: CHEBI:8984 SMILES: [Na+].CCCCCCCCCCCCOS([O-])(=O)=O
SDS Solution, 10% Sodium Dodecyl Sulfate Solution, Molecular Biology/Electrophoresis, Fisher BioReagents™
CAS: 151-21-3 Molecular Formula: C12H25NaO4S Molecular Weight (g/mol): 288.38 MDL Number: MFCD00036175 InChI Key: DBMJMQXJHONAFJ-UHFFFAOYSA-M PubChem CID: 3423265 ChEBI: CHEBI:8984 SMILES: [Na+].CCCCCCCCCCCCOS([O-])(=O)=O
Sodium Dodecyl Sulfate (Certified), Fisher Chemical
CAS: 151-21-3 Molecular Formula: C12H25NaO4S Molecular Weight (g/mol): 288.38 MDL Number: MFCD00036175 InChI Key: DBMJMQXJHONAFJ-UHFFFAOYSA-M Synonym: Sodium Lauryl Sulfate,SDS,Sulfuric Acid Monododecylester Sodium Salt,Irium PubChem CID: 3423265 ChEBI: CHEBI:8984 IUPAC Name: sodium dodecyl sulfate SMILES: [Na+].CCCCCCCCCCCCOS([O-])(=O)=O
Thermo Scientific Chemicals Dodecyl sulfate, sodium salt, 99%, for biochemistry
CAS: 151-21-3 Molecular Formula: C12H25NaO4S
Sodium Dodecylsulfate, 10% Solution, Ultrapure
CAS: 151-21-3 Molecular Formula: C12H25NaO4S Molecular Weight (g/mol): 288.38 MDL Number: MFCD00036175 InChI Key: DBMJMQXJHONAFJ-UHFFFAOYSA-M PubChem CID: 3423265 ChEBI: CHEBI:8984 SMILES: [Na+].CCCCCCCCCCCCOS([O-])(=O)=O
Thermo Scientific Chemicals Dodecyl sulfate, sodium salt, 98%, for biochemistry, suitable for electrophoresis
CAS: 151-21-3 | C12H25NaO4S | 288.38 g/mol
Dodecyl sulfate, sodium salt, 85%, pure
CAS: 151-21-3 Molecular Formula: C12H25NaO4S Molecular Weight (g/mol): 288.38 MDL Number: MFCD00036175 InChI Key: DBMJMQXJHONAFJ-UHFFFAOYSA-M Synonym: Dodecyl Sodium Sulfate,SDS,Sodium lauryl sulfate PubChem CID: 3423265 ChEBI: CHEBI:8984 IUPAC Name: sodium dodecyl sulfate SMILES: [Na+].CCCCCCCCCCCCOS([O-])(=O)=O
Sodium dodecyl sulfate, Suitable for electrophoresis, For molecular biology (GC), ≥99.0% (GC), MilliporeSigma™ Supelco™
CAS: 151-21-3 Molecular Weight (g/mol): 288.38 MDL Number: MFCD00036175 Synonym: Dodecyl sodium sulfate; Dodecyl sulfate sodium salt; Lauryl sulfate sodium salt; SDS; Sodium lauryl sulfate
Thermo Scientific Chemicals Dodecyl sulfate, sodium salt, 99+%, for molecular biology, DNAse, RNAse and Protease free
CAS: 151-21-3 | C12H25NaO4S | 288.38 g/mol