missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Dodecyl sulfate, sodium salt, 98%, for biochemistry, suitable for electrophoresis

Catalog No. AC419530050
Change view
Click to view available options
5kg; Plastic drum
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AC419530050 5kg; Plastic drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC419530050 Supplier Thermo Scientific™ Supplier No. 419530050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

For biochemistry

Specifications

Chemical Name or Material Dodecyl sulfate, sodium salt
CAS 151-21-3, 7757-82-6, 7647-14-5
Quantity 5kg
Assay Percent Range 97.5% min. (GC)
Molecular Formula C12H25NaO4S
Linear Formula CH3(CH2)11OSO3Na
MDL Number MFCD00036175
Beilstein 01, III, 1786
Flash Point >150°C
Fieser 13,281
Merck Index 15, 8768
Synonym sodium dodecyl sulfate, sodium lauryl sulfate, sodium dodecylsulfate, sodium lauryl sulphate, sodium dodecyl sulphate, dodecyl sulfate, sodium salt, anticerumen, gardinol, neutrazyme, duponal
Solubility Information Solubility in water: 130g/L (20°C). Other solubilities: partly soluble in alcohol, soluble 75g/L in ethanol (20°C)
InChI Key DBMJMQXJHONAFJ-UHFFFAOYSA-M
SMILES [Na+].CCCCCCCCCCCCOS([O-])(=O)=O
IUPAC Name sodium;dodecyl sulfate
Molecular Weight (g/mol) 288.38
PubChem CID 3423265
ChEBI CHEBI:8984
Formula Weight 288.38
Percent Purity 98%
Grade Biochemistry
Infrared Spectrum Authentic
Loss on Drying 1% max.
Packaging Plastic drum
Show More Show Less

RUO – Research Use Only