missing translation for 'onlineSavingsMsg'
Learn More

Sodium Dodecyl Sulfate (SDS), ≥95%

Catalog No. AAJ18220A7
Change view
Click to view available options
Quantity:
500 g
1 kg
5 kg
Packaging:
Cardboard Carton
Plastic bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AAJ18220A7 5 kg Cardboard Carton
AAJ1822036 500 g Plastic bottle
AAJ18220A1 1 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ18220A7 Supplier Thermo Fisher Scientific Supplier No. J18220A7

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 151-21-3 | C12H25NaO4S | 288.38 g/mol

Chemical Identifiers

CAS 151-21-3
Molecular Formula C12H25NaO4S
Molecular Weight (g/mol) 288.38
MDL Number MFCD00036175
InChI Key DBMJMQXJHONAFJ-UHFFFAOYSA-M
Synonym Sodium Lauryl Sulfate,n to Dodecyl sulfate sodium salt
PubChem CID 3423265
ChEBI CHEBI:8984
SMILES [Na+].CCCCCCCCCCCCOS([O-])(=O)=O

Specifications

Physical Form Crystalline Powder
Melting Point ∼206°C
Quantity 5 kg
Packaging Cardboard Carton
Linear Formula CH3(CH2)11OSO3Na
UN Number UN2926
Beilstein 3599286
IUPAC Name sodium;dodecyl sulfate
Formula Weight 288.38
Percent Purity ≥95%
Chemical Name or Material Sodium Dodecyl Sulfate (SDS)
Show More Show Less
Hazard Category H302-H315-H318-H500
Precautionary Statement P264b-P270-P280-P301+P312-P302+P352-P305+P351+P338-P310-P330-P332+P313-P362-P501c
EINECSNumber 205-788-1
RTECSNumber WT1050000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only