missing translation for 'onlineSavingsMsg'
Learn More

Sodium Dodecyl Sulfate (Certified), Fisher Chemical

Catalog No. O267425
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O267425 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. O267425 Supplier Thermo Fisher Scientific Supplier No. O2674 25

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 151-21-3
Molecular Formula C12H25NaO4S
Molecular Weight (g/mol) 288.38
MDL Number MFCD00036175
InChI Key DBMJMQXJHONAFJ-UHFFFAOYSA-M
Synonym Sodium Lauryl Sulfate, SDS, Sulfuric Acid Monododecylester Sodium Salt, Irium
PubChem CID 3423265
ChEBI CHEBI:8984
IUPAC Name sodium dodecyl sulfate
SMILES [Na+].CCCCCCCCCCCCOS([O-])(=O)=O

Specifications

Absorbance Pass Test
Color White
Physical Form Solid
Melting Point 206°C
pH 8.5 to 10
Quantity 25 g
Flash Point 100°C
Identification Pass Test
Packaging Glass Bottle
Assay Percent Range 95 to 105%
Linear Formula CH3(CH2)11OSO3Na
UN Number 1325
Formula Weight 288.38
Percent Purity 95 to 105%
Grade Certified
Chemical Name or Material Sodium Dodecyl Sulfate
Show More Show Less
Safety handling Check Alert storage code Red
Hazard Category DANGER!
Hazard Statement
Harmful if swallowed
Causes skin irritation
Causes serious eye damage
Precautionary Statement Emergency Overview
General Advice: If symptoms persist, call a physician.
Eye Contact: Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Get medical attention.
Skin Contact: Wash off immediately with plenty of water for at least 15 minutes. If skin irritation persists, call a physician.
Inhalation: Remove to fresh air. If not breathing, give artificial respiration. Get medical attention if symptoms occur.
Ingestion: Clean mouth with water and drink afterwards plenty of water. Get medical attention if
symptoms occur.
Most important symptoms and effects: Causes eye burns. Causes severe eye damage.

Prevention
Wash face, hands and any exposed skin thoroughly after handling. Do not eat, drink or smoke when using this product. Wear protective gloves/protective clothing/eye protection/face protection.

NFPA
Health: 3
Flammability: 1
Instability: 1
DOTInformation Class 4.1: Flammable Solid