missing translation for 'onlineSavingsMsg'
Learn More

SDS Solution, 20% Sodium Dodecyl Sulfate Solution, Molecular Biology/Electrophoresis, Fisher BioReagents™

Catalog No. BP13111
Change view
Click to view available options
Quantity:
200 mL
1 L
Packaging:
Nalgene Poly Bottle
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
BP13111 1 L Nalgene Poly Bottle
BP1311200 200 mL Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BP13111 Supplier Fisher BioReagents Supplier No. BP1311 1
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 151-21-3 | Molecular Formula: C12H25NaO4S | Molecular Weight: 288.38 g/mol

SDS Solution, 20% Sodium Dodecyl Sulfate Solution, Molecular Biology/Electrophoresis

  • DNase, RNase, and protease-free premixed solution containing 20% SDS for protein extraction and DNA extraction procedures
  • Reduce preparation time by using this premade SDS solution
  • SDS is a commonly used detergent in protein purification, polyacrylamide gel electrophoresis (PAGE), and other molecular biology applications
  • SDS is also used to disrupt cell membranes and dissociate nucleic acid-protein complexes in DNA extraction procedures
  • Intended for research use only; not for human or animal diagnostic purposes

Chemical Identifiers

CAS 151-21-3 , 7732-18-5
Molecular Formula C12H25NaO4S
Molecular Weight (g/mol) 288.38
MDL Number MFCD00036175
InChI Key DBMJMQXJHONAFJ-UHFFFAOYSA-M
Synonym Sodium lauryl sulfate, SDS
PubChem CID 3423265
ChEBI CHEBI:8984
SMILES [Na+].CCCCCCCCCCCCOS([O-])(=O)=O

Specifications

Absorbance 0.4 max. (0.1% solution) at 230nm, 0.1 max. (0.1% solution) at 280nm
Color Colorless
Physical Form Liquid
Boiling Point 100°C
Density 1.01g/cm³
Concentration 19.5 to 20.5%
Quantity 1 L
Identification Pass Test
Packaging Nalgene Poly Bottle
Assay Percent Range (as Fatty Alcohol Sulfate): ≥99%
Linear Formula CH3(CH2)10CH2OSO3Na
Merck Index 15, 8768
IUPAC Name sodium dodecyl sulfate
Formula Weight 288.38
Percent Purity ≥99%
Grade DNase-, RNase- and Protease-Free
Chemical Name or Material Sodium Dodecyl Sulfate, 20% Solution
Show More Show Less
Safety handling Check Alert storage code Red
Recommended Storage RT