missing translation for 'onlineSavingsMsg'
Learn More

Sodium Lauryl Sulfate, Pharmaceutical Secondary Standard, Certified Reference Material, MilliporeSigma™ Supelco™

Catalog No. 111018856
Change view
Click to view available options
Quantity:
2 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111018856 2 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111018856 Supplier Merck Emd Millipore Supplier No. PHR19492G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Specifications

CAS 151-21-3
Linear Formula CH3(CH2)11 OSO3Na
MDL Number MFCD00036175
Melting Point 204°C to 207°C
Synonym Sodium dodecyl sulfate; Dodecyl sodium sulfate; Dodecyl sulfate sodium salt; Lauryl sulfate sodium salt; SDS; Sodium lauryl sulfate
Quantity 2 g
Formula Weight 288.38
Grade Certified Reference Material