accessibility menu, dialog, popup

UserName

Uncover New Spotlight Deals and Get Up to 50% Off

Save Today

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Main Menu
Documents and Certificates

CAS RN 13820-91-2

NH3ClPtClClK

CAS RN 13820-91-2

IUPAC Name: potassium trichloroplatinumuide amine
Synonyms: potassium trichloroplatinumuide amine
Molecular Weight (g/mol): 357.56
Molecular Formula: Cl3H3KNPt
InChi Key: PIFYEDQIQOTXLO-UHFFFAOYSA-K
SMILES: N.[K+].Cl[Pt-](Cl)Cl
Quantity 
  • (1)
  • (1)
Grade 
  • (1)
Physical Form 
  • (1)

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.