missing translation for 'onlineSavingsMsg'
Learn More

Potassium trichloroammineplatinate(II)

CAS: 13820-91-2 | Cl3H3KNPt | 357.563 g/mol

Supplier:  Thermo Scientific Chemicals 039433ME

Catalog No. AA39433ME


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Potassium trichloroammineplatinate(II)
Crystalline
Cl3H3KNPt
potassium amminetrichloroplatinate ii, potassium trichloroammineplatinate ii, potassium trichloroplatinumuide amine, magnus red anion salt, potassium aminetrichloroplatinate ii, potassium;azane;trichloroplatinum 1-
N.Cl[Pt-](Cl)Cl.[K+]
357.563
357.56
13820-91-2
500 mg
MFCD00145154
PIFYEDQIQOTXLO-UHFFFAOYSA-K
potassium;azane;trichloroplatinum(1-)
11810450

Chemical Identifiers

13820-91-2
357.563
PIFYEDQIQOTXLO-UHFFFAOYSA-K
11810450
N.Cl[Pt-](Cl)Cl.[K+]
Cl3H3KNPt
MFCD00145154
potassium amminetrichloroplatinate ii, potassium trichloroammineplatinate ii, potassium trichloroplatinumuide amine, magnus red anion salt, potassium aminetrichloroplatinate ii, potassium;azane;trichloroplatinum 1-
potassium;azane;trichloroplatinum(1-)
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

RTECSNumber : TP1370000

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only