CAS RN 121643-44-5
CAS RN 121643-44-5
IUPAC Name:
2-methoxy-3-(trifluoromethyl)pyridine
Synonyms:
2-methoxy-3-(trifluoromethyl)pyridine
Molecular Weight (g/mol):
177.13
Molecular Formula:
C7H6F3NO
InChi Key:
SSAZZVQVJJXPMB-UHFFFAOYSA-N
SMILES:
COC1=NC=CC=C1C(F)(F)F
1
–
1
of
1
results
1
2-Methoxy-3-(trifluoromethyl)pyridine 98.0+%, TCI Americaâ„¢
CAS: 121643-44-5 Molecular Formula: C7H6F3NO Molecular Weight (g/mol): 177.13 MDL Number: MFCD00153202 InChI Key: SSAZZVQVJJXPMB-UHFFFAOYSA-N Synonym: 2-methoxy-3-trifluoromethyl pyridine,2-methoxy-3-trifluoromethyl-pyridine,pyridine,2-methoxy-3-trifluoromethyl,pyridine, 2-methoxy-3-trifluoromethyl,pubchem2404,acmc-209aes,tpc-py090,3-trifluoromethyl-2-methoxypyridine PubChem CID: 2775309 IUPAC Name: 2-methoxy-3-(trifluoromethyl)pyridine SMILES: COC1=NC=CC=C1C(F)(F)F