missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Methoxy-3-(trifluoromethyl)pyridine 98.0+%, TCI America™

$59.25 - $192.56

Chemical Identifiers

CAS 121643-44-5
Molecular Formula C7H6F3NO
Molecular Weight (g/mol) 177.126
MDL Number MFCD00153202
InChI Key SSAZZVQVJJXPMB-UHFFFAOYSA-N
Synonym 2-methoxy-3-trifluoromethyl pyridine, 2-methoxy-3-trifluoromethyl-pyridine, 3-trifluoromethyl-2-methoxypyridine, acmc-209aes, pubchem2404, pyridine, 2-methoxy-3-trifluoromethyl, pyridine,2-methoxy-3-trifluoromethyl, tpc-py090
PubChem CID 2775309
IUPAC Name 2-methoxy-3-(trifluoromethyl)pyridine
SMILES COC1=C(C=CC=N1)C(F)(F)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M21911G View Documents Tci America
M21911G
1g Each for $59.25
 
M21915G View Documents Tci America
M21915G
5g Each for $192.56
 
Specifications

Chemical Identifiers

121643-44-5
177.126
SSAZZVQVJJXPMB-UHFFFAOYSA-N
2775309
COC1=C(C=CC=N1)C(F)(F)F
C7H6F3NO
MFCD00153202
2-methoxy-3-trifluoromethyl pyridine, 2-methoxy-3-trifluoromethyl-pyridine, 3-trifluoromethyl-2-methoxypyridine, acmc-209aes, pubchem2404, pyridine, 2-methoxy-3-trifluoromethyl, pyridine,2-methoxy-3-trifluoromethyl, tpc-py090
2-methoxy-3-(trifluoromethyl)pyridine

Specifications

Colorless
121643-44-5
MFCD00153202
2-methoxy-3-trifluoromethyl pyridine, 2-methoxy-3-trifluoromethyl-pyridine, 3-trifluoromethyl-2-methoxypyridine, acmc-209aes, pubchem2404, pyridine, 2-methoxy-3-trifluoromethyl, pyridine,2-methoxy-3-trifluoromethyl, tpc-py090
COC1=C(C=CC=N1)C(F)(F)F
177.126
177.13
≥98.0% (GC)
1g
C7H6F3NO
1993
SSAZZVQVJJXPMB-UHFFFAOYSA-N
2-methoxy-3-(trifluoromethyl)pyridine
2775309
Clear Liquid at 20°C
2-Methoxy-3-(trifluoromethyl)pyridine
Safety and Handling

Safety and Handling

TSCA : No