Learn More
Zirconium sponge, 0.8-25.4mm (0.03-1.0in), 99.5%, Zr & Hf, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 01044118
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Gray | |
7440-67-7 | |
6.506g/cm3 | |
Odorless | |
Sponge | |
MFCD00011303 MFCD00148878 | |
QCWXUUIWCKQGHC-UHFFFAOYSA-N | |
zirconium | |
23995 |
Zirconium | |
Gray sponge | |
50 g | |
99.5% | |
Zr | |
zircat, powder, circonio, zirconium, elemental, zirconio, zirkonium, compounds, unii-c6v6s92n3c, and compounds, foil | |
[Zr] | |
91.22 | |
CHEBI:33342 |
Chemical Identifiers
7440-67-7 | |
91.22 | |
QCWXUUIWCKQGHC-UHFFFAOYSA-N | |
23995 | |
zirconium |
Zr | |
MFCD00011303 MFCD00148878 | |
zircat, powder, circonio, zirconium, elemental, zirconio, zirkonium, compounds, unii-c6v6s92n3c, and compounds, foilShow More | |
CHEBI:33342 | |
[Zr] |
Safety and Handling
P210-P222-P223-P231+P232-P280-P335+P334-P370+P378q-P501c
H250-H260
DOTInformation : DOT Class: 4.2; Packaging Group: II
EINECSNumber : 231-176-9
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only