missing translation for 'onlineSavingsMsg'
Learn More

Zeolite, type: ZSM-5

CAS: 308081-08-5 | C14H16N2 | 212.296 g/mol

$107.99 - $274.43

Chemical Identifiers

CAS 308081-08-5
Molecular Formula C14H16N2
Molecular Weight (g/mol) 212.296
MDL Number MFCD00132601
InChI Key HSWPZIDYAHLZDD-UHFFFAOYSA-N
Synonym atipamezole, antisedan, 4-2-ethyl-2-indanyl imidazole, 4-2-ethyl-2,3-dihydro-1h-inden-2-yl-1h-imidazole, atipamezol spanish, atipamezolum latin, unii-03n9u5jaf6, 1h-imidazole, 4-2-ethyl-2,3-dihydro-1h-inden-2-yl, 5-2-ethyl-2,3-dihydro-1h-inden-2-yl-1h-imidazole, dsstox_cid_29061Show MoreShow Less
PubChem CID 71310
IUPAC Name 5-(2-ethyl-1,3-dihydroinden-2-yl)-1H-imidazole
SMILES CCC1(CC2=CC=CC=C2C1)C3=CN=CN3
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC279571000
SDS View Documents
Thermo Scientific Chemicals
279571000
100 g
Each for $107.99
Only null left
 
AC279575000
SDS View Documents
Thermo Scientific Chemicals
279575000
500 g
Each for $274.43
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Adsorbent
Specifications
Show More
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves

GHS Signal Word: Warning

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only