Learn More
Labcon North America ZAP™ SLIK 1000uL Low Retention Aerosol Filter Pipet Tips, in Racks, Sterile
Supplier: Labcon North America 11779650089
Tip, Pipet; Labcon; ZAP™ SLIK Low Retention Aerosol Filter; Fits most popular 1000uL pipettors; Low mounting and ejection forces less than 3kg; Ultra low fluid retention; Sterile; Patented robotically inserted endotoxin free filter; Protease, Human DNA, ATP, RNase/DNase and Endotoxin Free (non-pyrogenic); Polypropylene USP Class VI resin; Validated Sterilization (ISO 11137) for use in Research and Medical labs; Refillable autoclavable racks; 96 tips/rack; 6 racks/pack; 8 packs/case; 88.9mm H.; Made in USA
Description
- Environmental benefits include:
- Renewable Energy