Learn More
Fisherbrand™ Zahn Cup-Type Viscometers
Fisherbrand Zahn Cup-Type Viscometers are used for quick, accurate measurements of viscosity of liquids such as paint, lacquer, varnish, syrup, oil, etc.
 View more versions of this product
Catalog No. 15-348-50E
Only null left
Explore available promotions
Description
Description
  • Fisherbrand™ Zahn Cup-Type Viscometers are portable cups that measure the viscosity of a liquid in Zahn seconds (the amount of time required for a steady-stream to flow from the orifice at the bottom of the cup)
  • Measure viscosity of very thin to very heavy liquids
  • Ideal for unknown liquid experiments
  • Quick, accurate measurement
  • Made from ASTM™ Standard Stainless Steel
  • Electronically passivated for corrosion resistance
  • Can be sterilized by Eto/gamma radiation/autoclave
  • Calibration certificate, conversion charts and instructions included
Specifications
Specifications
Cup Type Viscometer No. 5
Corrosion Resistant Stainless Steel
0.53 cm
35 cm
1.5 oz.
460 to 1725 cSt
0.208 in.
13.75 in.
For quick measurement of the viscosity of very heavy mixtures
44 mL
Product Suggestions
Product Suggestions