missing translation for 'onlineSavingsMsg'
Learn More

Yttrium(III) hexafluoro-2,4-pentanedionate

CAS: 18911-76-7 | C15H3F18O6Y | 710.06 g/mol

$89.86 - $310.70

Chemical Identifiers

CAS 18911-76-7
Molecular Formula C15H3F18O6Y
Molecular Weight (g/mol) 710.06
MDL Number MFCD00064764
InChI Key AJVDSGQQIVVYRX-UHFFFAOYSA-N
Synonym yttrium hexafluoroacetylacetonate, z-1,1,1,5,5,5-hexafluoro-4-hydroxypent-3-en-2-one; yttrium
PubChem CID 50930692
SMILES [Y+3].FC(F)(F)C(=O)[CH-]C(=O)C(F)(F)F.FC(F)(F)C(=O)[CH-]C(=O)C(F)(F)F.FC(F)(F)C(=O)[CH-]C(=O)C(F)(F)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA1471703
View Documents
Thermo Scientific Chemicals
01471703
1 g
Each for $89.86
Only null left
 
AA1471706
View Documents
Thermo Scientific Chemicals
01471706
5 g
Each for $310.70
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

18911-76-7
710.06
AJVDSGQQIVVYRX-UHFFFAOYSA-N
50930692
C15H3F18O6Y
MFCD00064764
yttrium hexafluoroacetylacetonate, z-1,1,1,5,5,5-hexafluoro-4-hydroxypent-3-en-2-one; yttrium
[Y+3].FC(F)(F)C(=O)[CH-]C(=O)C(F)(F)F.FC(F)(F)C(=O)[CH-]C(=O)C(F)(F)F.FC(F)(F)C(=O)[CH-]C(=O)C(F)(F)F

Specifications

166°C to 170°C
Crystalline
C15H3F18O6Y
yttrium hexafluoroacetylacetonate, z-1,1,1,5,5,5-hexafluoro-4-hydroxypent-3-en-2-one; yttrium
[Y+3].FC(F)(F)C(=O)[CH-]C(=O)C(F)(F)F.FC(F)(F)C(=O)[CH-]C(=O)C(F)(F)F.FC(F)(F)C(=O)[CH-]C(=O)C(F)(F)F
710.06
710.1
18911-76-7
1 g
MFCD00064764
AJVDSGQQIVVYRX-UHFFFAOYSA-N
yttrium(3+) tris(1,1,1,5,5,5-hexafluoro-2,4-dioxopentan-3-ide)
50930692
Yttrium(III) hexafluoro-2,4-pentanedionate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only