Learn More
Yttrium(III) acetate tetrahydrate, 99.9% (REO)
In analytical chemistry | CAS: 85949-60-6 | C6H17O10Y | 338.10 g/mol
Supplier: Thermo Scientific Chemicals 01456518
Description
In analytical chemistry.Yttrium(III) acetate tetrahydrate is used in analytical chemistry and in catalysis. It is used in the preparation of yttrium trifluoride nanoparticles, which shows antibacterial properties.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Yttrium(III) acetate tetrahydrate | |
White | |
Powder | |
(REO) | |
Y(OOCCH3)3·4H2O | |
Soluble in hot water. | |
O.O.O.O.[Y+3].CC([O-])=O.CC([O-])=O.CC([O-])=O | |
338.10 | |
99.9% |
(decomposition) | |
85949-60-6 | |
50 g | |
C6H17O10Y | |
MFCD00013048 | |
AIQRTHPXPDTMBQ-UHFFFAOYSA-K | |
yttrium(3+) triacetate tetrahydrate | |
338.10 (266.04 Anhydrous) | |
1.5 g/mL |
Chemical Identifiers
85949-60-6 | |
338.10 | |
AIQRTHPXPDTMBQ-UHFFFAOYSA-K | |
O.O.O.O.[Y+3].CC([O-])=O.CC([O-])=O.CC([O-])=O |
C6H17O10Y | |
MFCD00013048 | |
yttrium(3+) triacetate tetrahydrate |
Safety and Handling
EINECSNumber : 245-612-0
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only