missing translation for 'onlineSavingsMsg'
Learn More

Ytterbium(III) chloride hexahydrate, 99.99+%, (trace metal basis)

CAS: 10035-01-5 | Cl3Yb·6H2O | 387.48 g/mol

Supplier:  Thermo Scientific Chemicals 437180500

 View more versions of this product

Catalog No. AC437180500

Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for ultra trace analysis
Specifications

Specifications

Ytterbium(III) chloride hexahydrate
Brown to Brown-Red
Crystals
For trace metal basis analysis
Cl3Yb·6H2O
Solubility in water: soluble. Other solubilities: soluble in absolute alcohol
387.48
Trace Metal Basis
865°C
10035-01-5
50 g
99.99+%
YbCl3·6H2O
387.48
≥99.99%
Glass bottle

Chemical Identifiers

10035-01-5
387.48
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.

GHS Signal Word: Warning

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only