missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Superior Uniform Worklon™ Reusable Polyester Taffeta Cleanroom Frocks
Taffeta can be used in a variety of industries requiring particulate control and resistance to chemicals and abrasion. Garments are gamma compatible and autoclavable.
Supplier: Superior Uniform 478S
Description
- Adjustable snaps at wrist and collar
- Color-coded hang loop denotes size
- ISO 6, Class 1000 cleanroom compatible
- Unisex design
- LD-100
Greener Choice Claims
- Environmental benefits include:
- Renewable Energy
Specifications
Controlled Environments Frock | |
Non-sterile | |
Mandarin | |
Class 1000 (ISO 6) |
Reusable | |
Snaps | |
Autoclavable |