missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Wheaton™ Lab 45 Graduated Bottles With Cap Attached

Graduated bottles with cap attached

Supplier:  DWK Life Sciences 219930

 View more versions of this product

Catalog No. 06-451-292


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

33.8 oz., 1000 mL
33.8 oz.
3.97 in.
Borosilicate Glass
Yes
233 mm
Polypropylene
101 mm
1000 mL
Clear
Pouring ring and screw cap
White
Wide
9.17 in.
45 mm
Screw Cap
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions