Learn More
DWK Life Sciences Wheaton™ Celstir™ Spinner Double Side Arm Flasks without Water Jacket
Provide maximum surface between culture and glass flask
Supplier:  DWK Life Sciences 356887
 View more versions of this product
Catalog No. 06-416-2F
Only null left
Explore available promotions
Description
Description
Provide maximum surface between culture and glass flask
  • PTFE-resin coated glass impeller does not protrude through cap and has bottom dimple for more efficient stirring
  • Silicone rubber-lined cap
  • Autoclavable
Greener Choice Claims
  • Environmental benefits include:
    • Recyclable
    • Certifications and Seals of Approval
Specifications
Specifications
Silicone Rubber
4.5 cm
13.4 in.
Flask, impeller and caps
Autoclavable
10 to 44 cm
7 in.
3000 mL
34.1 cm
PTFE Resin
17.8 cm
Product Suggestions
Product Suggestions