missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Wheaton™ Celstir™ Spinner Double Side Arm Flasks without Water Jacket

Provide maximum surface between culture and glass flask

Supplier:  DWK Life Sciences 356882

 View more versions of this product

Catalog No. 06-416-2E


Only null left
Explore available promotions

Description

Description

Provide maximum surface between culture and glass flask

  • PTFE-resin coated glass impeller does not protrude through cap and has bottom dimple for more efficient stirring
  • Silicone rubber-lined cap
  • Autoclavable

Greener Choice Claims
  • Environmental benefits include:
    • Recyclable
Specifications

Specifications

Silicone Rubber
4.5 cm
Borosilicate Glass
19 cm
PTFE Resin
11 cm
4.3 in.
500 mL
7.4 in.
Flask, impeller and caps
Autoclavable
10 to 44 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions