Learn More
Cytiva Whatman™ Type 4 Chr Chromatography Paper
Supplier:  Cytiva 30048098
Catalog No. 09-800-981
Only null left
Explore available promotions
Description
Description
Greener Choice Claims
  • Environmental benefits include:
    • Non-Toxic
    • Recyclable
    • Renewable Energy
Specifications
Specifications
20 mm x 100 mm
Recommended for Routine and/or Repetitive Chromatography When Loadings Are Relatively Low
180 mm/30 min.
0.21 mm
Cellulose
Product Suggestions
Product Suggestions