Learn More
Cytiva Whatman™ pH Indicator Papers, CF Strips Edge
Edge
Provides an extremely clear and accurate visual pH value using four different segments of dye-impregnated indicator papers. Whatman™ pH Indicator Papers, CF Strips use dyes that are chemically bonded to the paper and cannot be leached into solution.
Supplier:  Cytiva 2614991
 View more versions of this product
Catalog No. 09-876-18
Only null left
Edge
Explore available promotions
Description
Description
Provides an extremely clear and accurate visual pH value using four different segments of dye-impregnated indicator papers. Whatman™ pH Indicator Papers, CF Strips use dyes that are chemically bonded to the paper and cannot be leached into solution.
  • Plastic supports that retain stiffness in liquids
  • Paper segments on each strip have chemically bonded dyes; can't leach into solution
  • Complementary indicator charts provide clear comparison
  • Full-range strips (0 to 14) graduated in 1pH units; wide-range strips (4.5 to 10pH) in 0.5pH units
  • Packs of 100; each strip is approximately 0.25 Ă— 3 in. (6 Ă— 80mm)
Greener Choice Claims
  • Environmental benefits include:
    • Renewable Energy
Specifications
Specifications
4.5 to 10
0.5 pH
100 Pack
80 x 6 mm
6 mm
Strip
Plastic supports, paper segments on each strip, complementary indicator charts
pH Test Strip, Color Bonded
80 mm
Product Suggestions
Product Suggestions