missing translation for 'onlineSavingsMsg'
Learn More

Water, for residue analysis, ECD tested for pesticide analysis
GREENER_CHOICE

CAS: 7732-18-5 | H2O | 18.015 g/mol

$120.53 - $240.78

Chemical Identifiers

CAS 7732-18-5
Molecular Formula H2O
Molecular Weight (g/mol) 18.015
MDL Number MFCD00011332
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym dihydrogen oxide, dihydrogen monoxide
PubChem CID 962
ChEBI CHEBI:15377
IUPAC Name oxidane
SMILES O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC326650010
SDS View Documents
Thermo Scientific Chemicals
326650010
1 L Glass Bottle
Each for $120.53
Only null left
 
AC326650025
SDS View Documents
Thermo Scientific Chemicals
326650025
2.5 L Glass Bottle
Each for $240.78
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Show More
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 231-791-2

RTECSNumber : ZC0110000

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only