missing translation for 'onlineSavingsMsg'
Learn More

Water, Deionized, ASTM Type I, Certified ACS Grade, LabChem™

Supplier:  LabChem LC267405

 View more versions of this product

Catalog No. LC267405


Only null left

Description

Description

Specifications
Show More
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Product is not hazardous.

EINECSNumber : 231-791-2

Recommended Storage : Room Temperature

SDS
Documents

Documents

Promotions

Promotions