missing translation for 'onlineSavingsMsg'
Learn More

Water, 0.1% Formic Acid, filtered through a 0.2 micron filter, BAKER ANALYZED™ LC-MS Reagent, J.T. Baker™

Catalog No. p-5383338
Change view
Click to view available options
Quantity:
1 L
4 L
Packaging:
Narrow Mouth Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
02004566 1 L Narrow Mouth Amber Glass Bottle
02004568 4 L Narrow Mouth Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 02004566 Supplier Avantor Supplier No. 983402
Only null left

Function tested and optimized for LC/MS applications

    For use in liquid chromatography and mass spectrometry; For laboratory, research, or manufacturing use

Chemical Identifiers

CAS 7732-18-5
Molecular Formula H2O
Molecular Weight (g/mol) 18.015
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym water, dihydrogen oxide, distilled water, purified water, water vapor, sterile water, water, purified, water, deionized, water, mineral, dihydrogen monoxide
PubChem CID 962
ChEBI CHEBI:15377
IUPAC Name oxidane
SMILES O

Specifications

Grade BAKER ANALYZED™ LC-MS Reagent
Packaging Narrow Mouth Amber Glass Bottle
Quantity 1 L
Specific Gravity 1L = 1.00kg
Chemical Name or Material Water, 0.1% Formic Acid; filtered through a 0.2 micron filter