missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Wall-Mounted Stainless-Steel Shelves
Easily assembled employing slip-fit TAB-LOK design
$852.32 - $852.32
Specifications
Product Type | Wall Shelf |
---|---|
For Use With (Application) | Convenient storage space for spices, timers etc |
Catalog Number | Mfr. No. | Length (English) | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Length (English) | Price | Quantity | ||||||
19-033-595D
![]() |
Fisherbrand™
WSKD60 |
60 in. |
Each for $852.32
|
|
||||||
Description
- 18 gauge stainless-steel polished to satin finish
- Knocked down (KD) type shelf
- Die stamped and die formed shelf and brackets
- Furnished with 1.5 in. sanitary rolled rim with 1.25 in. turn-up edge on sides and rear
Specifications
Wall Shelf | |
Convenient storage space for spices, timers etc |