missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
SmartLine Vortex Mixer
The SmartLine Vortex Mixer features a dynamic balancing system and powerful motor to provide instant vortexing of even the largest sample sizes, including full 50mL tubes.
Manufacturer: SmartLine BV1000GMFIS
Description
- Unique balancing system of the mixer creates maximum vortexing action, while minimizing noise and excessive vibration
- Powerful for instant vortexing
- 200 to 3200 variable speed
- Continuous or “touch” operation
- Handy size (LxWxH): 16 x 13 x 17cm (6.3 x 5 x 6.7 in.)
Certifications
CE marked
Warranty
2 years
Specifications
115V, 60Hz |