missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Poltex Visual Management Mask Box Holder

Holds one box of masks in use, and 2 back up boxes
$286.75 - $324.49
Specifications
Description | Holds 3 boxes of masks: 1 in use, two back up |
---|---|
Dimensions (L x W x H) | 4.5 x 7.75 x 11 in. |
Description
- When open box is finished, lift front slider to remove empty box, full box falls into place
- Provides visual management of working stock
- Durable PETG construction
Specifications
Holds 3 boxes of masks: 1 in use, two back up | |
4.5 x 7.75 x 11 in. |