Learn More
SOMNI Scientific™ Vaporizer Service Exchange Iso Tec 5 (Specify Fill Type, Mount)
Maintains device accuracy, proper function and prevents exposure from leaks. A fully serviced and calibrated vaporizer of the same type is shipped to the customer. Customer uses prepaid return label to send their vaporizer back to our service center. No downtime.
Supplier: SOMNI Scientific™ VX6017
Description
- Isoflurane, Tec 5
- Specify Fill type, Mount and Serial number when ordering
- Maintains accuracy
- Prevents exposure from leaks
- Full Service and Calibration
- Certificate of Calibration included
- Service interval based on manufacturer recommendation
- Warranty provided
Greener Choice Claims
- Environmental benefits include:
- Renewable Energy
Specifications
RM-17003, RM-17004, AMD-17004, TT-17000, OC-21003, OC-21004 | |
16.5 cm | |
Animal Inhalant anesthesia | |
Mounting hardware, Calibration Certificate |
Brass and Alloys | |
10.8 cm | |
Tec 5 Isoflurane |
For Veterinary Use Only