Learn More
Honeywell™ Uvex Flex Seal™ Safety Goggles
GREENER_CHOICE
Honeywell ™ Uvex Flex Seal™ Safety Goggles with ANSI, and CSA safety compliance
$24.20 - $24.20
Specifications
Certifications/Compliance ANSI Z87+ (High Impact), ANSI Z.87.1/1989, CA 21,199, CSA Z94.3
Lens Tint Clear
Product Type Safety Eyewear
For Use With (Application) Abrasions, Airborne Particulate, Biohazard, Chemical, Impact, Splash
Frame Color Navy
View More Specs
Products 1
Catalog Number Mfr. No. Headband Material Price Quantity  
Catalog Number Mfr. No. Headband Material Price Quantity  
19085466
View Documents
Honeywell™
S3400HS
Neoprene
Each for $24.20
Case of 50 Each for $1,137.50
Only null left
Description
Description
  • Lightweight, silicone body conforms to the face assuring the highest level of comfort and eye protection
  • Hydroshield™ AF coating provides 2x greater scratch resistance than other Honeywell™ anti-fog coatings
  • Uvextreme AF lens coating offers consistent fog-free time 90x longer than the majority of other anti-fog products
  • Tested by Honeywell™ to meet the requirements of the CSA Z94.3 standard
  • Silicone body stays supple even in extreme temperatures
  • Exceptional chemical splash and impact resistance
  • Fits over most prescription eyewear
  • Indirect venting system
  • Made in the USA
Specifications
Specifications
ANSI Z87+ (High Impact), ANSI Z.87.1/1989, CA 21,199, CSA Z94.3
Safety Eyewear
Navy
50/Pk.
Universal
0.3 kg
Silicone
Clear
Abrasions, Airborne Particulate, Biohazard, Chemical, Impact, Splash
Headband, Anti-Fog lens
1 Ea.
Indirect
Polycarbonate
Uvextreme AF Anti-Fog Coating
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'