Learn More
Honeywell™ Uvex™ Protege™ Safety Eyewear with Hydroshield™ Anti-Fog Lens Coating
Delivers anti-fog protection that lasts up to 60X longer
Supplier:  Honeywell™ S4206HS
 View more versions of this product
Catalog No. 19-185-759
Only null left
Explore available promotions
Description
Description
  • HydroShield™ anti-fog lens coating
  • Provides 99.9% ultra-violet protection
  • Provides 2X greater scratch-resistance
Compliance
Meet ANSI™ Z87.1-2010 standards. Meet military V0 standard for impact protection.
Specifications
Specifications
Anti-fog
HydroShield AF
SCT-Low IR
Black
Product Suggestions
Product Suggestions