Learn More
BIOTIX™ uTIP™ Filter Sterile Low Retention Universal Pipette Tips, Standard
GREENER_CHOICE
Experience a better universal tip. Biotix™ uTIP™ Filter Sterile Pipette Tips made with Flexfit™ creates a secure seal on various pipettes with less insertion and ejection force.
$237.33 - $1186.70
Specifications
Filtered Filtered
Sterility Sterile
Compatibility Universal
Product Type Pipette Tips
Disposable Single-use
View More Specs
Products 7
Catalog Number Mfr. No. Volume (Metric) Price Quantity  
Catalog Number Mfr. No. Volume (Metric) Price Quantity  
12111000
GREENER_CHOICE
View Documents Promotion Details
BIOTIX™
63300154
10 μL
Pack of 960 Each for $237.33
Case of 5 Pack for $1,186.65
Only null left
12111002
GREENER_CHOICE
View Documents Promotion Details
BIOTIX™
63300042
20 μL
Pack of 960 Each for $237.33
Case of 5 Pack for $1,186.65
Only null left
12111003
GREENER_CHOICE
View Documents Promotion Details
BIOTIX™
63300043
100 μL
Pack of 960 Each for $237.33
Case of 5 Pack for $1,186.65
Only null left
12111004
GREENER_CHOICE
View Documents Promotion Details
BIOTIX™
63300045
300 μL
Pack of 960 Each for $237.33
Case of 5 Pack for $1,186.65
Only null left
12111005
GREENER_CHOICE
View Documents Promotion Details
BIOTIX™
63300047
1250 μL
Pack of 960 Each for $296.67
Case of 4 Pack for $1,186.68
Only null left
12-111-126
GREENER_CHOICE
View Documents Promotion Details
BIOTIX™
63300044
200 μL
Case of 4800 Each for $1,186.70
Only null left
12-111-132
GREENER_CHOICE
View Documents Promotion Details
BIOTIX™
63300046
1000 μL
Case of 3840 Each for $1,186.70
Only null left
Description
Description
Experience a better universal tip with Biotix™ uTIP™
  • Biotix filter sterile tips offers aerosol protection to prevent cross contamination
  • Fits all standard universal shaft single channel, multichannel and electronic pipettes
  • FlexFit technology features flexible proximal end to ensure secure seal with 66 to 80% less insertion force than competitor tips
  • X-Resin drastically reduces sample retention resulting in tighter CV values
  • Blade technology lowers chance of hanging droplet formation by optimizing surface area at distal end of tip
  • Each lot of Biotix tips are tested and certified free of RNase, DNase, Endotoxins/Pyrogens, PCR Inhibitors, trace metals, and human DNA
Certifications
ISO 9001
Specifications
Specifications
Filtered
Universal
Single-use
Yes
Polypropylene
Sterile
Pipette Tips
Universal Fit
Biotix uTip
Conical
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'