missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Urea (Colorless-to-White Crystals or Crystalline Powder/Mol. Biol.), Fisher BioReagents™
$107.45 - $1,639.93

Chemical Identifiers
CAS | 57-13-6 |
---|---|
Molecular Formula | CH4N2O |
Molecular Weight (g/mol) | 60.056 |
InChI Key | XSQUKJJJFZCRTK-UHFFFAOYSA-N |
Synonym | alphadrate, carbamide, carbamimidic acid, carbonyldiamide, carbonyldiamine, isourea, pseudourea, ureaphil, ureophil, urevert |
PubChem CID | 1176 |
ChEBI | CHEBI:48376 |
IUPAC Name | urea |
SMILES | C(=O)(N)N |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
BP169-500
|
Fisher BioReagents
BP169500 |
500g | Poly Bottle |
Each for $107.45
Case of 6 Each for $644.70 |
|
BP169-212
|
Fisher BioReagents
FLBP169212 |
2.5kg | Poly Pail |
Each for $409.10
|
|
BP169-10
|
Fisher BioReagents
BP16910 |
10kg | Poly Drum |
Each for $1,639.93
|
|
Description
Urea is used as a denaturing agent for DNA and proteins.Chemical Identifiers
57-13-6 | |
60.056 | |
alphadrate, carbamide, carbamimidic acid, carbonyldiamide, carbonyldiamine, isourea, pseudourea, ureaphil, ureophil, urevert | |
CHEBI:48376 | |
C(=O)(N)N |
CH4N2O | |
XSQUKJJJFZCRTK-UHFFFAOYSA-N | |
1176 | |
urea |
Specifications
57-13-6 | |
7.5 to 9.5 | |
131°C | |
≥99 % | |
NH2CONH2 | |
15, 10052 | |
500g | |
C(=O)(N)N | |
60.056 | |
CHEBI:48376 | |
Solid | |
DNase-, RNase- and Protease-Free | |
Urea |
0.05 max. (5M solution) at 280nm | |
White | |
Pass Test | |
CH4N2O | |
Poly Bottle | |
alphadrate, carbamide, carbamimidic acid, carbonyldiamide, carbonyldiamine, isourea, pseudourea, ureaphil, ureophil, urevert | |
XSQUKJJJFZCRTK-UHFFFAOYSA-N | |
urea | |
1176 | |
60.06 | |
≥99% | |
1.25mmHg at 25°C |
Safety and Handling

Recommended Storage : RT